Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Calpeptin, ≥98%

Catalog No. p-7082234
Click to view available options
Quantity:
10 mg

Inhibitor of the calcium-dependent protease calpain and cathepsin L

Calpeptin is used to to inhibit the activities of cathepsin L and calpain-1. Calpeptin blocked the expression of aspartyl protease cathepsin D, amyloid precursor protein, and calparin. It acts as an inhibitor of the Ca2+-dependent protease, calpain. Prevents collagen- and thrombin-induced platelet aggregation, probably by blocking calpain induced phospholipase C and thromboxane synthase activation. Potent cathepsin L inhibitor. It is shown to preferentially inhibit a subset of protein-tyrosine phosphatases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117591-20-5
Molecular Formula C20H30N2O4, C20H30N2O4
Synonym N-Benzyloxycarbonyl-L-leucyl-norleucinal; Z-Leu-norleucinal

Specifications

Quantity 10 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) Inhibitor of the calcium-dependent protease calpain and cathepsin L
Formula Weight 362.50g/mol
Solubility Soluble in DMSO at 15mg/ml
Product Type Signal Transduction Reagent
Form Powder
InChI Key PGGUOGKHUUUWAF-ROUUACIJSA-N
SMILES CCCC[C@H](NC(=O)[C@H](CC(C)C)NC(=O)OCC1=CC=CC=C1)C=O
IUPAC Name benzyl N-[(1S)-3-methyl-1-{[(2S)-1-oxohexan-2-yl]carbamoyl}butyl]carbamate
Molecular Weight (g/mol) 362.47
Assay ≥98%
Show More Show Less
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.