Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CalPlus™ (Brand of Calcium Chloride Dihydrate), Multi-Compendial, U.S.P., J.T. Baker™

Catalog No. p-5390732 Shop All Avantor Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
Wide Mouth PVC Coated Glass Bottle
This item is not returnable. View return policy

Catalog No. 02-002-521

Supplier: Avantor 133601

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Suitable for Use in Hemodialysis

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·2H2O

Specifications

Quantity 500 g
Formula Weight 147.01
Grade Multi-Compendial/USP
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 0.84kg
Chemical Name or Material CalPlus™ (Brand of Calcium Chloride Dihydrate)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.