Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calponin 1 Rabbit anti-Human, Clone: CNN1/4227R, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP30758920UG

 View more versions of this product

Catalog No. NB122652


Only null left
Add to Cart

Description

Description

Calponin 1 Monoclonal specifically detects Calponin 1 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Calponin 1
Monoclonal
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
Basic calponin, calponin 1, basic, smooth muscle, Calponin H1, smooth muscle, calponins, basic, Sm-Calp, SMCCcalponin-1
Synthetic peptide corresponding to residues on the C-terminus of human Calponin 1 was used as an immunogen.
20 μg
Cancer, Cell Biology, Cellular Markers
1264
Store at 4°C.
IgG
Immunohistochemistry (Paraffin)
CNN1/4227R
Unconjugated
10 mM PBS with 0.05% BSA
Rabbit
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.