Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calponin 1 Rabbit anti-Human, Clone: CNN1/4227R, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$304.50

Specifications

Antigen Calponin 1
Clone CNN1/4227R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122652
SDP
View Documents
Novus Biologicals
NBP30758920UG
20 μg
Each of 1 for $304.50
Only null left
Add to Cart
 
Description

Description

Calponin 1 Monoclonal specifically detects Calponin 1 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Calponin 1
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Cell Biology, Cellular Markers
10 mM PBS with 0.05% BSA
1264
IgG
Protein A or G purified
CNN1/4227R
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human
Basic calponin, calponin 1, basic, smooth muscle, Calponin H1, smooth muscle, calponins, basic, Sm-Calp, SMCCcalponin-1
Synthetic peptide corresponding to residues on the C-terminus of human Calponin 1 was used as an immunogen.
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.