Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calponin Mouse anti-Bovine, Human, Porcine, Unconjugated, Clone: CALP or N3/16, MilliporeSigma™
Mouse Monoclonal Antibody
$456.56
Specifications
Antigen | Calponin |
---|---|
Clone | CALP or N3/16 |
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABT1504-100
|
MilliporeSigma
MABT1504100UG |
100 μg |
Each of 1 for $456.56
|
|
|||||
Description
Anti-Calponin, clone CALP , Cat. No. MABT1504, is a mouse monoclonal antibody that detects Calponin-1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.Specifications
Calponin | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Bovine, Human, Pig | |
Basic calponin;Calponin H1 smooth muscle | |
CNN1 | |
IgG1 κ | |
Protein G purified | |
Clone CALP specifically detects human Calponin-1. | |
∽36 kDa observed; 33.17 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
CALP or N3/16 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cell Structure | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_001290 | |
Crude human uterine muscle extract. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
Antibodies |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title