Learn More
Calponin Mouse anti-Human, Clone: 1A11E5, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
| Antigen | Calponin |
|---|---|
| Clone | 1A11E5 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6960
![]()
|
Proteintech Group Inc
665401IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody raised against the specific region of human CNN1, thus it does not cross-react with CNN2 or CNN3.
Calponin, a thin filament associated protein is implicated in the regulation and modulation of smooth muscle contraction. It is capable of binding to actin, calmodulin, troponin C and tropomyosin. Calponin is expressed in smooth muscle and tissues containing significant amounts of smooth muscle. Two isoforms of calponin exist whose molecular weights are 34kDa and 29kDa. Expression of the 29kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and maligt breast lesions. It stains smooth muscle, myoepithelial cells, myofibroblasts, keratinocytes and nerve fibers. It identifies myoepithelial cells in breast lesions, and helps differentiate breast collagenous spherulosis (positive) from adenoid cystic carcinoma. Adenoid cystic carcinoma in salivary gland tumors is calponin positive.Specifications
| Calponin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Basic calponin, Calponin 1, Calponin H1, smooth muscle, CNN1, Sm Calp, SMCC | |
| CNN1 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 1A11E5 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P51911 | |
| 1264 | |
| Calponin Fusion Protein Ag21384 | |
| Primary | |
| -20°C | |
| CNN1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.