Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calreticulin-2/CALR3 Antibody (321007), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB304210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304210 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304210 Supplier Novus Biologicals Supplier No. FAB2927N

Mouse Monoclonal Antibody

Calreticulin-2/CALR3 Monoclonal antibody specifically detects Calreticulin-2/CALR3 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Calreticulin-2/CALR3
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 321007
Conjugate Alexa Fluor 700
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias calreticulin 2, calreticulin 3, Calreticulin-2, calreticulin-3, cancer/testis antigen 93, CRT2calreticulin-2, CT93, FLJ25355, MGC26577
Host Species Mouse
Immunogen E. coli-derived recombinant human Calreticulin-2, Thr20-Leu384, Accession # Q96L12
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 125972
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.