Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calreticulin-2/CALR3 Antibody (OTI1E1), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP272380G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272380G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272380G Supplier Novus Biologicals Supplier No. NBP272380G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calreticulin-2/CALR3 Monoclonal specifically detects Calreticulin-2/CALR3 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot.

Specifications

Antigen Calreticulin-2/CALR3
Applications Western Blot
Classification Monoclonal
Clone OTI1E1
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias calreticulin 2, calreticulin 3, Calreticulin-2, calreticulin-3, cancer/testis antigen 93, CRT2calreticulin-2, CT93, FLJ25355, MGC26577
Gene Symbols CALR3
Host Species Mouse
Immunogen Full length recombinant protein of human Calreticulin-2/CALR3 (NP_659483) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 125972.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.