Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calreticulin Antibody (1G6A7), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB147518AFB Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB147518AFB 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB147518AFB Supplier Novus Biologicals Supplier No. NBP147518AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calreticulin Monoclonal specifically detects Calreticulin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Calreticulin
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1G6A7
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias autoantigen Ro, calregulin, calreticulin, cC1qR, CRP55, CRT, CRTC, Endoplasmic reticulum resident protein 60, ERp60, FLJ26680, grp60, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSAHACBP
Gene Symbols CALR
Host Species Mouse
Immunogen Calreticulin Antibody (1G6A7) was developed against a synthetic peptide corresponding to the C-terminus (EEEDVPGQAKDELC) of human Calreticulin, conjugated to KLH. [UniProt# P27797]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cellular Markers, ER Markers, Immunology, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 811
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.