Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calreticulin Antibody (681233), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB120471 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120471 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120471 Supplier Novus Biologicals Supplier No. FAB38981Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calreticulin Monoclonal specifically detects Calreticulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunohistochemistry, Western Blot.

Specifications

Antigen Calreticulin
Applications Flow Cytometry (Intracellular Staining), Gene Knock-Out, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 681233
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias autoantigen Ro, CALR, calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA
Host Species Mouse
Immunogen E. coli-derived recombinant human Calreticulin, Glu18-Leu417, Accession # P27797
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cellular Markers, ER Markers, Immunology, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 811
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.