Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calreticulin Antibody (6C6), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB236319 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB236319 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB236319 Supplier Novus Biologicals Supplier No. NBP250053AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calreticulin Monoclonal antibody specifically detects Calreticulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Calreticulin
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 6C6
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias autoantigen Ro, CALR, calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA
Host Species Mouse
Immunogen Calreticulin Antibody (6C6) was developed against a synthetic peptide VESGSLEDDWDFLPPKKI corresponding to amino acids 191-208 of human Calreticulin, including the LC3 interacting region or LIR. [UniProt# P27797]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cellular Markers, ER Markers, Immunology, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 811
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.