Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calreticulin Antibody (CR213-2AG), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB120570 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120570 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120570 Supplier Novus Biologicals Supplier No. NBP311672JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calreticulin Monoclonal specifically detects Calreticulin in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Calreticulin
Applications Western Blot, ELISA
Classification Monoclonal
Clone CR213-2AG
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias autoantigen Ro, CALR, calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA
Host Species Mouse
Immunogen Recombinant human calreticulin.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cellular Markers, ER Markers, Immunology, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 811
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.