Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calsenilin Antibody (011), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB072007 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB072007 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB072007 Supplier Novus Biologicals Supplier No. NBP290302AF405
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Calsenilin Monoclonal specifically detects Calsenilin in Human samples. It is validated for ELISA.

Specifications

Antigen Calsenilin
Applications ELISA
Classification Monoclonal
Clone 11
Conjugate Alexa Fluor 405
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias calsenilin, calsenilin, presenilin-binding protein, EF hand transcription factor, CSEN MGC18289, DREAM, DREAMA-type potassium channel modulatory protein 3, KChIP3, KCHIP3 DRE-antagonist modulator, Kv channel interacting protein 3, calsenilin, Kv channel-interacting protein 3, potassium channel interacting protein 3
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Calsenilin (Q9Y2W7-1; Met1-Ile256).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 30818
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.