Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calsenilin Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB065738 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB065738 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB065738 Supplier Novus Biologicals Supplier No. NBP300359UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calsenilin Polyclonal specifically detects Calsenilin in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Calsenilin
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias calsenilin, calsenilin, presenilin-binding protein, EF hand transcription factor, CSEN MGC18289, DREAM, DREAMA-type potassium channel modulatory protein 3, KChIP3, KCHIP3 DRE-antagonist modulator, Kv channel interacting protein 3, calsenilin, Kv channel-interacting protein 3, potassium channel interacting protein 3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Calsenilin (Q9Y2W7-1; Met1-Ile256).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 30818
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.