Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calsequestrin 2 Antibody (4E10), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP174043A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP174043A30 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP174043A30 Supplier Novus Biologicals Supplier No. NBP174043AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calsequestrin 2 Monoclonal specifically detects Calsequestrin 2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Calsequestrin 2
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 40000000000
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias calsequestrin 2 (cardiac muscle), calsequestrin 2, fast-twitch, cardiac muscle, Calsequestrin, cardiac muscle isoform, calsequestrin-2, FLJ26321, FLJ93514, PDIB2
Gene Symbols CASQ2
Host Species Mouse
Immunogen Recombinant human CASQ2 (20-399aa) purified from E. coli
Quantity 100 μL
Research Discipline Cardiovascular Biology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 845
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.