Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calsequestrin 2 Antibody (4E10), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP174043R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP174043R 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP174043R Supplier Novus Biologicals Supplier No. NBP174043R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calsequestrin 2 Monoclonal specifically detects Calsequestrin 2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Calsequestrin 2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 40000000000
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias calsequestrin 2 (cardiac muscle), calsequestrin 2, fast-twitch, cardiac muscle, Calsequestrin, cardiac muscle isoform, calsequestrin-2, FLJ26321, FLJ93514, PDIB2
Gene Symbols CASQ2
Host Species Mouse
Immunogen Recombinant human CASQ2 (20-399aa) purified from E. coli
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cardiovascular Biology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 845
Test Specificity Anti-human CASQ2 mAb, clone 4E10, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human CASQ2 protein.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.