Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaM Kinase I Antibody (010), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB334839 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334839 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334839 Supplier Novus Biologicals Supplier No. NBP290023MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CaM Kinase I Monoclonal antibody specifically detects CaM Kinase I in Human samples. It is validated for ELISA

Specifications

Antigen CaM Kinase I
Applications ELISA
Classification Monoclonal
Clone 010
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias calcium/calmodulin-dependent protein kinase I, calcium/calmodulin-dependent protein kinase type 1, CaM kinase I, CaM kinase I alpha, caM-KI, CaMKI, CaMKI-alpha, EC 2.7.11, EC 2.7.11.17, MGC120317, MGC120318
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human CaM Kinase I (Accession#: NP_003647.1, Leu2-Leu370)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Protein Kinase, Sensory Systems, Vision
Primary or Secondary Primary
Gene ID (Entrez) 8536
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.