Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaM Kinase II alpha Antibody (6G9), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB1001983R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001983R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001983R Supplier Novus Biologicals Supplier No. NB1001983R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CaM Kinase II alpha Monoclonal specifically detects CaM Kinase II alpha in Bovine, Human, Mouse, Rat, Xenopus species samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, KnockDown.

Specifications

Antigen CaM Kinase II alpha
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, KnockDown
Classification Monoclonal
Clone 6G9
Conjugate DyLight 550
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P11275
Gene Alias calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, CaM kinase II alpha subunit, CaM kinase II subunit alpha, CAMKAcalcium/calmodulin-dependent protein kinase type II subunit alpha, CaMK-II alpha subunit, CaMK-II subunit alpha, CaMKIINalpha, CaM-kinase II alpha chain, EC 2.7.11, EC 2.7.11.17, KIAA0968calcium/calmodulin-dependent protein kinase type II alpha chain
Gene Symbols CAMK2A
Host Species Mouse
Immunogen Partially purified rat CaMKII
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Neuroscience, Neurotransmission, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 815
Test Specificity Detects approx 50-60kDa. Recognizes both phosphorylated and non-phosphorylated forms.
Target Species Bovine, Human, Mouse, Rat, Xenopus species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.