Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaM Kinase II alpha Antibody (JM11-07), Novus Biologicals™
SDP

Catalog No. NBP267207 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267207 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267207 Supplier Novus Biologicals Supplier No. NBP267207
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CaM Kinase II alpha Monoclonal specifically detects CaM Kinase II alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen CaMKinaseIIalpha
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JM11-07
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Flow Cytometry 1:50-1:100, Immunoprecipitation 1:10-1:50, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, CaM kinase II alpha subunit, CaM kinase II subunit alpha, CAMKAcalcium/calmodulin-dependent protein kinase type II subunit alpha, CaMK-II alpha subunit, CaMK-II subunit alpha, CaMKIINalpha, CaM-kinase II alpha chain, EC 2.7.11, EC 2.7.11.17, KIAA0968calcium/calmodulin-dependent protein kinase type II alpha chain
Gene Symbols CAMK2A
Host Species Rabbit
Immunogen Synthetic peptide within Human CaM Kinase II alpha aa 37-71/ 478. (SwissProt: Q9UQM7 Human; SwissProt: P11798 Mouse; SwissProt: P11275 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Neuroscience, Neurotransmission, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 815
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.