Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaM Kinase II alpha, p Thr286 Antibody (SR1853), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322608100UL
Description
CaM Kinase II alpha Monoclonal antibody specifically detects CaM Kinase II alpha in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, SerumSpecifications
CaM Kinase II alpha | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR1853 | |
Western Blot 1:500-1:2000, Immunohistochemistry, Serum 1:50-1:200 | |
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, CaM kinase II alpha subunit, CaM kinase II subunit alpha, CAMKAcalcium/calmodulin-dependent protein kinase type II subunit alpha, CaMK-II alpha subunit, CaMK-II subunit alpha, CaMKIINalpha, CaM-kinase II alpha chain, EC 2.7.11, EC 2.7.11.17, KIAA0968calcium/calmodulin-dependent protein kinase type II alpha chain | |
A synthesized peptide derived from human phospho-CaM Kinase II alpha [Thr286] (Uniprot #: Q9UQM7, P11798, P11275) | |
100 μg | |
Neuroscience, Neurotransmission, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases, Wnt Signaling Pathway | |
815 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction