Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaM Kinase II alpha, p Thr286 Antibody (SR1853), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | CaM Kinase II alpha |
---|---|
Clone | SR1853 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry, Serum 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
CaM Kinase II alpha Monoclonal antibody specifically detects CaM Kinase II alpha in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, SerumSpecifications
CaM Kinase II alpha | |
Western Blot 1:500-1:2000, Immunohistochemistry, Serum 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Rat | |
calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, CaM kinase II alpha subunit, CaM kinase II subunit alpha, CAMKAcalcium/calmodulin-dependent protein kinase type II subunit alpha, CaMK-II alpha subunit, CaMK-II subunit alpha, CaMKIINalpha, CaM-kinase II alpha chain, EC 2.7.11, EC 2.7.11.17, KIAA0968calcium/calmodulin-dependent protein kinase type II alpha chain | |
A synthesized peptide derived from human phospho-CaM Kinase II alpha [Thr286] (Uniprot #: Q9UQM7, P11798, P11275) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1853 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Neuroscience, Neurotransmission, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases, Wnt Signaling Pathway | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
815 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title