Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaM Kinase II Antibody (SR1856), Novus Biologicals™
SDP

Catalog No. NB387895 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387895 50 μL
NB259871 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB387895 Supplier Novus Biologicals Supplier No. NBP32244750UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CaM Kinase II Monoclonal antibody specifically detects CaM Kinase II in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen CaM Kinase II
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone SR1856
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias Calcium/calmodulin-dependent protein kinase;Calcium/calmodulin-dependent protein kinase type II subunit beta;CAM2;CAMK2;CAMK2B;CAMKB;CaMKIIbeta;MRD54
Host Species Rabbit
Immunogen A synthesized peptide derived from human CaM Kinase II (Uniprot number: Q9UQM7, Q13554, Q13555, Q13557)
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Cell Biology, Cell Cycle and Replication, GPCR, Immunology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 816
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.