Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaM Kinase II Antibody (SR1956), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321943100UL
Description
CaM Kinase II Monoclonal antibody specifically detects CaM Kinase II in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
CaM Kinase II | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
SR1956 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Calcium/calmodulin-dependent protein kinase;Calcium/calmodulin-dependent protein kinase type II subunit beta;CAM2;CAMK2;CAMK2B;CAMKB;CaMKIIbeta;MRD54 | |
A synthesized peptide derived from human CaM Kinase II (Uniprot #: Q9UQM7, Q13554, Q13555, Q13557) | |
100 μg | |
Apoptosis, B Cell Development and Differentiation Markers, Cell Biology, Cell Cycle and Replication, GPCR, Immunology, Neuroscience, Signal Transduction | |
816 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction