Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaM Kinase II Antibody (SR1956), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | CaM Kinase II |
---|---|
Clone | SR1956 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
CaM Kinase II Monoclonal antibody specifically detects CaM Kinase II in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
CaM Kinase II | |
Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Calcium/calmodulin-dependent protein kinase;Calcium/calmodulin-dependent protein kinase type II subunit beta;CAM2;CAMK2;CAMK2B;CAMKB;CaMKIIbeta;MRD54 | |
A synthesized peptide derived from human CaM Kinase II (Uniprot #: Q9UQM7, Q13554, Q13555, Q13557) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1956 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Apoptosis, B Cell Development and Differentiation Markers, Cell Biology, Cell Cycle and Replication, GPCR, Immunology, Neuroscience, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
816 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title