Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaM Kinase II delta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CaM Kinase II delta |
---|---|
Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CaM Kinase II delta Polyclonal specifically detects CaM Kinase II delta in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CaM Kinase II delta | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
calcium/calmodulin-dependent protein kinase (CaM kinase) II delta, calcium/calmodulin-dependent protein kinase II delta, calcium/calmodulin-dependent protein kinase type II delta chain, calcium/calmodulin-dependent protein kinase type II subunit delta, CaM kinase II delta subunit, CaM kinase II subunit delta, CAMKD, CaMK-II delta subunit, CaMK-II subunit delta, CaM-kinase II delta chain, DKFZp686G23119, DKFZp686I2288, EC 2.7.11, EC 2.7.11.17, MGC44911 | |
CAMK2D | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Rabbit | |
Protein Kinase, Wnt Signaling Pathway | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
817 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MDGSGMPKTMQSEETRVWHRRDGKWQNVHFHRSGSPTVPIKPPCIPNGKENFSGGTSLWQ | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title