Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaM Kinase II delta Rabbit anti-Human, Mouse, Rat, Clone: 10M1P2, Novus Biologicals™
SDP

Catalog No. NB168658 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168658 20 μg
NB168657 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168658 Supplier Novus Biologicals Supplier No. NBP31678320UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CaM Kinase II delta Monoclonal antibody specifically detects CaM Kinase II delta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen CaM Kinase II delta
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 10M1P2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias calcium/calmodulin-dependent protein kinase (CaM kinase) II delta, calcium/calmodulin-dependent protein kinase II delta, calcium/calmodulin-dependent protein kinase type II delta chain, calcium/calmodulin-dependent protein kinase type II subunit delta, CaM kinase II delta subunit, CaM kinase II subunit delta, CAMKD, CaMK-II delta subunit, CaMK-II subunit delta, CaM-kinase II delta chain, DKFZp686G23119, DKFZp686I2288, EC 2.7.11, EC 2.7.11.17, MGC44911
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CaM Kinase II delta (Q13557).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 817
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.