Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CaM Kinase II gamma Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CaM Kinase II gamma |
---|---|
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CaM Kinase II gamma Polyclonal specifically detects CaM Kinase II gamma in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
CaM Kinase II gamma | |
Polyclonal | |
Rabbit | |
Protein Kinase, Signal Transduction, Tyrosine Kinases, Wnt Signaling Pathway | |
calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma, calcium/calmodulin-dependent protein kinase II gamma, calcium/calmodulin-dependent protein kinase type II subunit gamma, CaM kinase II subunit gamma, CAMK, CAMKGFLJ16043, CAMK-II, CaMK-II subunit gamma, EC 2.7.11, EC 2.7.11.17, MGC26678 | |
CAMK2G | |
IgG | |
Affinity Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
818 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NSLVSPAQEPAPLQTAMEPQTTVVHNATDGIKGSTESCNTTTEDEDLKAAPLRTGNGSSVPEGRSSR | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title