Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMKID Antibody (OTI2C6), FITC, Novus Biologicals™
SDP

Catalog No. NBP272074F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272074F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272074F Supplier Novus Biologicals Supplier No. NBP272074F

Mouse Monoclonal Antibody

CAMKID Monoclonal specifically detects CAMKID in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CAMKID
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI2C6
Conjugate FITC
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias calcium/calmodulin-dependent protein kinase ID, calcium/calmodulin-dependent protein kinase type 1D, CaM kinase I delta, CaM kinase ID, CaM-K1, CaMKI delta, CaM-KI delta, CaMKID, CaMKI-like protein kinase, CKLiKEC 2.7.11.17, EC 2.7.11
Gene Symbols CAMK1D
Host Species Mouse
Immunogen Full length recombinant protein of human CAMKID (NP_065130) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57118.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.