Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaMKII alpha/beta, p Thr286, p Thr287 Antibody (22B1), Biotin, Novus Biologicals™
SDP

Catalog No. NB11096869B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11096869B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11096869B Supplier Novus Biologicals Supplier No. NB11096869B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CaMKII alpha/beta Monoclonal specifically detects CaMKII alpha/beta in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.

Specifications

Antigen CaMKII alpha/beta (p Thr286)
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot
Classification Monoclonal
Clone 22B1
Conjugate Biotin
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P11275
Gene Alias calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, CaM kinase II alpha subunit, CaM kinase II subunit alpha, CAMKAcalcium/calmodulin-dependent protein kinase type II subunit alpha, CaMK-II alpha subunit, CaMK-II subunit alpha, CaMKIINalpha, CaM-kinase II alpha chain, EC 2.7.11, EC 2.7.11.17, KIAA0968calcium/calmodulin-dependent protein kinase type II alpha chain
Gene Symbols CAMK2A
Host Species Mouse
Immunogen Synthetic peptide
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 815
Test Specificity This antibody is specific for a and B subunits of CaMKII only when they are phosphorylated at Thr-286/287 (in B).
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.