Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaMKIIbeta/gamma Rabbit anti-Human, Clone: SU03-57, Novus Biologicals™
SDP

Catalog No. NBP267476 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267476 Supplier Novus Biologicals Supplier No. NBP267476

Rabbit Monoclonal Antibody

CaMKII beta/gamma Monoclonal specifically detects CaMKII beta/gamma in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CaMKIIbeta/gamma
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SU03-57
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias calcium/calmodulin-dependent protein kinase (CaM kinase) II beta, calcium/calmodulin-dependent protein kinase II beta, calcium/calmodulin-dependent protein kinase type II beta chain, calcium/calmodulin-dependent protein kinase type II subunit beta, CaM kinase II beta subunit, CaM kinase II subunit beta, CAM2CAMK2, CAMKB, CaMK-II subunit beta, CaM-kinase II beta chain, EC 2.7.11, EC 2.7.11.17, MGC29528, proline rich calmodulin-dependent protein kinase
Gene Symbols CAMK2B
Host Species Rabbit
Immunogen Synthetic peptide within Human CaMKII beta/gamma aa 201-250/ 478. (SwissProt: Q13554 Human; SwissProt: Q13555 Human; SwissProt: Q13557 Human; SwissProt: Q9UQM7 Human; SwissProt: P11798 Mouse; SwissProt: P28652 Mouse; SwissProt: Q6PHZ2 Mouse; SwissProt: Q923T9 Mouse; SwissProt: P08413 Rat; SwissProt: P11275 Rat; SwissProt: P11730 Rat; SwissProt: P15791 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Protein Kinase, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 816
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.