Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaMKIV Antibody (001), Biotin, Novus Biologicals™
SDP

Catalog No. NB040888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB040888 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB040888 Supplier Novus Biologicals Supplier No. NBP290733B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CaMKIV Monoclonal specifically detects CaMKIV in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CaMKIV
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1
Conjugate Biotin
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias brain Ca(2+)-calmodulin-dependent protein kinase type IV, brain Ca++-calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase IV, calcium/calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase type IV catalytic chain, CAM kinase- GR, CAM kinase IV, CaM kinase-GR, caMK, CaMK IV, CaMK-GR, EC 2.7.11, EC 2.7.11.17, IV, MGC36771
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CaMKIV (P08414; Met1-Tyr469).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 814
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.