Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaMKIV Antibody, Novus Biologicals™
SDP

Catalog No. NB1203557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1203557 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1203557 Supplier Novus Biologicals Supplier No. NB1203557
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CaMKIV Polyclonal specifically detects CaMKIV in Human, Mouse, Rat, Canine, Plant, Primate, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen CaMKIV
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias brain Ca(2+)-calmodulin-dependent protein kinase type IV, brain Ca++-calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase IV, calcium/calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase type IV catalytic chain, CAM kinase- GR, CAM kinase IV, CaM kinase-GR, caMK, CaMK IV, CaMK-GR, EC 2.7.11, EC 2.7.11.17, IV, MGC36771
Gene Symbols CAMK4
Host Species Rabbit
Immunogen Synthetic Peptide: V(127) E K G Y Y S E R D A A D A V K Q(143) C
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 814
Test Specificity NB120-3557 detects CaM kinase IV (CaMKIV) from human, mouse, rat, canine, primate, arabidopsis and Xenopus Oocyte samples.
Target Species Human, Mouse, Rat, Canine, Plant, Primate, Xenopus species
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.