Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaMKV Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB181283 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB181283 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB181283 Supplier Novus Biologicals Supplier No. NBP306485F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CaMKV Polyclonal antibody specifically detects CaMKV in Human samples. It is validated for Immunofluorescence

Specifications

Antigen CaMKV
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias 1G5, CaM kinase-like vesicle-associated, caM kinase-like vesicle-associated protein, MGC8407, VACAMKL, vesicle-associated calmodulin-binding protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CaMKV fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 79012
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.