Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAMP/Cathelicidin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications
$207.00 - $499.50
Specifications
Concentration | LYOPH |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Antisera |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017938
![]() |
Novus Biologicals
NB100986890.025ML |
0.025 mL |
Each for $207.00
|
|
|||||
NB10098689
![]() |
Novus Biologicals
NB10098689 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
CAMP/Cathelicidin Polyclonal specifically detects CAMP/Cathelicidin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.Specifications
LYOPH | |
Unconjugated | |
Rabbit | |
Mouse | |
820 | |
IgG | |
Unpurified | |
Specific for Cathelin related antimicrobial peptide. |
Polyclonal | |
Antisera | |
RUO | |
P51437 | |
CAMP | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title