Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMP/Cathelicidin Antibody, Novus Biologicals™
SDP

Catalog No. NB017938 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017938 0.025 mL
NB10098689 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017938 Supplier Novus Biologicals Supplier No. NB100986890.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

CAMP/Cathelicidin Polyclonal specifically detects CAMP/Cathelicidin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.

Specifications

Antigen CAMP/LL37/FALL39/Cathelicidin
Applications Immunohistochemistry, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:2000, Immunohistochemistry-Frozen
Formulation Whole antisera with No Preservative
Gene Accession No. P51437
Gene Alias 18 kDa cationic antimicrobial protein, CAP18CRAMP, cathelicidin antimicrobial peptide, FALL-39, FALL39CAP-18, hCAP-18, LL37
Gene Symbols CAMP
Host Species Rabbit
Immunogen A synthetic peptide from mouse Cathelin related antimicrobial peptide (Cathelicidin, CAMP, CRAMP) conjugated to an immunogenic carrier protein has been used as the antigen.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 820
Test Specificity Specific for Cathelin related antimicrobial peptide.
Reconstitution Reconstitute with 0.1 ml sterilized water to desired concentration. Add glycerol (1:1) for additional stability.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.