Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMP/Cathelicidin Antibody (OSX12), Novus Biologicals™
SDP

Catalog No. NBP146781SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146781SS 0.025 mg
NBP146781 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP146781SS Supplier Novus Biologicals Supplier No. NBP146781SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CAMP/Cathelicidin Monoclonal specifically detects CAMP/Cathelicidin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen CAMP/Cathelicidin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone OSX12
Conjugate Unconjugated
Dilution Western Blot 5-10 ug/ml, Flow Cytometry 5-10 ug/ml, Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence 5-10 ug/ml, Immunoprecipitation, Immunohistochemistry-Paraffin 5-10 ug/ml
Formulation Sterilized water with No Preservative
Gene Accession No. P49913
Gene Alias 18 kDa cationic antimicrobial protein, Antibacterial peptide FALL-39, CAP18, CAP-18, cathelicidin antimicrobial peptide, CRAMP, epididymis secretory sperm binding protein, FALL39, FALL-39, FALL-39 peptide antibiotic, hCAP-181, HSD26, LL37
Gene Symbols CAMP
Host Species Mouse
Immunogen A synthetic peptide from human CAMP/Cathelicidin conjugated to an immunogenic carrier protein has been used as the antigen.
Purification Method Ammonium sulfate precipitation
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 820
Test Specificity Specific for Cathelicidin antimicrobial peptide.
Reconstitution Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.