Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMP/LL37/FALL39/Cathelicidin Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N176864J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N176864J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N176864J646 Supplier Novus Biologicals Supplier No. NBP176864JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CAMP/LL37/FALL39/Cathelicidin Polyclonal specifically detects CAMP/LL37/FALL39/Cathelicidin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CAMP/LL37/FALL39/Cathelicidin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 18 kDa cationic antimicrobial protein, CAP18CRAMP, cathelicidin antimicrobial peptide, FALL-39, FALL39CAP-18, hCAP-18, LL37
Gene Symbols CAMP
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from an internal portion of the human Cathelicidin protein. The immunogen is located within amino acids 50 - 100 of Cathelicidin.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 820.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.