Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMP/LL37/FALL39/Cathelicidin Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB338092 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338092 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338092 Supplier Novus Biologicals Supplier No. NBP176864MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CAMP/LL37/FALL39/Cathelicidin Polyclonal antibody specifically detects CAMP/LL37/FALL39/Cathelicidin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CAMP/LL37/FALL39/Cathelicidin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 18 kDa cationic antimicrobial protein, CAP18CRAMP, cathelicidin antimicrobial peptide, FALL-39, FALL39CAP-18, hCAP-18, LL37
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from an internal portion of the human Cathelicidin protein. The immunogen is located within amino acids 50 - 100 of Cathelicidin.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 820
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.