Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-Camphene (contains ca. 20% Tricyclene) 78.0+%, TCI America™
SDP

Catalog No. C000925G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g

Catalog No. C000925G

Supplier: TCI America C000925G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79-92-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00066603
InChI Key CRPUJAZIXJMDBK-UHFFFAOYSA-N
Synonym camphene, comphene, 2,2-dimethyl-3-methylenenorbornane, 3,3-dimethyl-2-methylenenorbornane, 3,3-dimethyl-2-methylenenorcamphane, +/--camphene, 2,2-dimethyl-3-methylenebicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylidenebicyclo 2.2.1 heptane, bicyclo 2.2.1 heptane, 2,2-dimethyl-3-methylene, fema no. 2229
PubChem CID 6616
ChEBI CHEBI:3830
IUPAC Name 3,3-dimethyl-2-methylidenebicyclo[2.2.1]heptane
SMILES CC1(C2CCC(C2)C1=C)C

Specifications

Melting Point 46°C
Color White-Yellow
Boiling Point 159°C
Quantity 25 g
UN Number 1325
Formula Weight 136.24
Percent Purity ≥78.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (+/-)-Camphene (contains ca. 20% Tricyclene)
EINECSNumber (4)-0613
RTECSNumber EX1055000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.