Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Campylobacter Jejuni Antibody (380/412), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB162426AG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162426AG 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB162426AG Supplier Novus Biologicals Supplier No. NB10062426AF488

Mouse Monoclonal Antibody

Campylobacter Jejuni Monoclonal specifically detects Campylobacter Jejuni in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Campylobacter Jejuni
Applications ELISA
Classification Monoclonal
Clone 380/412
Conjugate Alexa Fluor 488
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Campilobacter
Host Species Mouse
Immunogen C. jejuni Penner 2
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-62426 is specific for Campylobacter jejuni. Some reactivity has been observed with Penner 4 +/- and with Penner 10. C. jejuni is a curved, rod-shaped gram negative bacterium which is a member of the Campylobacteraceae family. They are motile with a polar unsheathed flagellum that results in a corkscrew like motion. They occur primarily as commensals in humans and domestic animals, particularly in chickens where gut temperatures of 42oC are optimal for growth. C. jejuni are one of the most common causes of human diarrhea in the world. Food poisoning can be severely debilitating, although rarely life-threatening as the disease is self limiting. Campylobacter infections are also associated with postinfectious complications including arthritis, Reiter syndrome and GuillainûBarre┤ syndrome.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.