Learn More
Campylobacter Jejuni Mouse anti-Bacteria, Clone: 2E-10., Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Campylobacter Jejuni |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 2E-10. |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA 1 - 5 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml |
| Formulation | 10mM PBS with 0.05% BSA with 0.05% Sodium Azide |
| Gene Alias | Campilobacter |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.