Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Campylobacter Venerealis Antibody (1053/350) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064517
Description
Campylobacter Venerealis Monoclonal specifically detects Campylobacter Venerealis in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Campylobacter Venerealis | |
Monoclonal | |
1.0 mg/mL | |
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Mouse | |
Protein A purified | |
RUO | |
BTGs applied: EC, CAM, ENT 1, 2, 3, 4, W. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG3 |
ELISA, Immunocytochemistry, Immunofluorescence | |
1053/350 | |
Unconjugated | |
Vibrio fetus | |
Campylobacter fetus (NCTC 10842, ATCC 27374). | |
0.1 mg | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction