Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Campylobacter Venerealis Antibody (1053/350) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Campylobacter Venerealis |
---|---|
Clone | 1053/350 |
Concentration | 1.0 mg/mL |
Dilution | ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
Applications | Immunofluorescence, Immunocytochemistry, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017593
![]() |
Novus Biologicals
NB100645170.025MG |
0.025 mg |
Each for $181.90
|
|
|||||
NB10064517
![]() |
Novus Biologicals
NB10064517 |
0.1 mg |
Each for $487.50
|
|
|||||
Description
Campylobacter Venerealis Monoclonal specifically detects Campylobacter Venerealis in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Campylobacter Venerealis | |
1.0 mg/mL | |
Immunofluorescence, Immunocytochemistry, ELISA | |
Unconjugated | |
Mouse | |
Bacteria | |
IgG3 | |
Protein A purified | |
BTGs applied: EC, CAM, ENT 1, 2, 3, 4, W. |
1053/350 | |
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Campylobacter fetus (NCTC 10842, ATCC 27374). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title