Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Campylobacter Venerealis Antibody (1053/350), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB10064517U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064517U 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064517U Supplier Novus Biologicals Supplier No. NB10064517UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Campylobacter Venerealis Monoclonal specifically detects Campylobacter Venerealis in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Campylobacter Venerealis
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1053/350
Conjugate DyLight 350
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Gene Alias Vibrio fetus
Host Species Mouse
Immunogen Campylobacter fetus (NCTC 10842, ATCC 27374).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity BTGs applied: EC, CAM, ENT 1, 2, 3, 4, W.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.