Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMSAP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP126645 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP126645 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP126645 Supplier Novus Biologicals Supplier No. NBP126645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

CAMSAP1 Polyclonal specifically detects CAMSAP1 in Human, Rat samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen CAMSAP1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 3-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_056262.3
Gene Alias bA100C15.1, bA100C15.1 (novel protein), calmodulin regulated spectrin-associated protein 1, calmodulin-regulated spectrin-associated protein 1, DKFZp434F195, DKFZp434G2311, FLJ31228, MGC163452, PRO2405
Gene Symbols CAMSAP1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1552 and 1602 of human calmodulin regulated spectrin-associated protein 1 using the numbering given in entry NP_056262.3 (GeneID 157922).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157922
Test Specificity Rat reactivity reported in scientific literature (PMID: 24486153). Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, Chimpanzee, White-tufted-ear marmoset and Northern white-cheeked gibbon.
Target Species Human, Rat
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.