Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAMSAP1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | CAMSAP1 |
---|---|
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
CAMSAP1 Polyclonal antibody specifically detects CAMSAP1 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
CAMSAP1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
bA100C15.1, bA100C15.1 (novel protein), calmodulin regulated spectrin-associated protein 1, calmodulin-regulated spectrin-associated protein 1, DKFZp434F195, DKFZp434G2311, FLJ31228, MGC163452, PRO2405 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 350-650 of human CAMSAP1 (NP_056262.3). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Stem Cells | |
PBS, 50% glycerol, pH7.3 | |
157922 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title