Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMSAP1L1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP121402 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP121402 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP121402 Supplier Novus Biologicals Supplier No. NBP121402
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

CAMSAP1L1 Polyclonal specifically detects CAMSAP1L1 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockdown Validated.

Specifications

Antigen CAMSAP1L1
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, KnockDown
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q08AD1
Gene Alias calmodulin regulated spectrin-associated protein 1-like 1, calmodulin-regulated spectrin-associated protein 2, CAMSAP2, KIAA1078Calmodulin-regulated spectrin-associated protein 1-like protein 1, MGC150680, MGC150681
Gene Symbols CAMSAP2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1439 and 1489 of human calmodulin regulated spectrin-associated protein 1-like 1 using the numbering given in entry EAW91320.1 (GeneID 23271).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23271
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Crab-eating macaque, African elephant, Chinese hamster, Small-eared galago and Northern white-cheeked gibbon. Rat reactivity reported in scientific literature (PMID: 24440982).
Target Species Human, Rat
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.