Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMSAP1L1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501561930
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-1930 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-1930 Supplier Bethyl Laboratories, Inc Supplier No. A302162AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

CAMSAP2 gene ontology annotations related to this gene include neuron projection development.

Specifications

Antigen CAMSAP1L1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CAMSAP2
Gene Accession No. Q08AD1
Gene Alias calmodulin regulated spectrin-associated protein 1-like 1, calmodulin regulated spectrin-associated protein family, member 2, calmodulin-regulated spectrin-associated protein 1-like protein 1, CAMSAP1L1, KIAA1078
Gene Symbols CAMSAP2
Host Species Rabbit
Immunogen Between 1439 and 1489
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23271
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.