Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cancer Diagnostics, Inc.™ Tissue Marking Dyes
SDP

Catalog No. 22500019 Shop All Cancer Diagnostics Products
Click to view available options
Quantity:
2 oz.
8 oz.
Color:
Black
Blue
Green
Green, Black, Red, Yellow, Blue, Orange
Green, Black, Red, Yellow, Blue, Orange, Violet
Orange
Red
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Formulated to be used without a mordant

  • Dyes are pigment-based with an alcohol foundation for consistency and durability
  • Proprietary blend of materials remains bright through processing and produces clean, crisp results
  • Will not run or bleed on tissue
  • Impervious to laboratory chemicals and reagents

Order Info

Individual colors available in two sizes; also available in systems with sets of six or seven colors, wood tray and 50 applicator sticks. Two-year shelf life.

Specifications

Boiling Point 93°C
Color Blue
Product Type Tissue Marking Dye
Quantity 8 oz.
Formulation Isopropyl Alcohol 400ppm, Ammonia 35ppm
pH 8.5 to 10
Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.