Learn More
Cancer Diagnostics, Inc.™ Cytology Staining Reagents
SDP
Catalog No. 22500049 Shop All Cancer Diagnostics Products
Click to view available options
Type:
EA-50 Cytology Counter Stain
EA-50 Modified Cytology Counter Stain
EA-65 Cytology Counter Stain
OG-6 Cytology Stain
This item is not returnable. View return policy
This item is not returnable. View return policy
For use in routine cytoplasmic staining
EA-50, EA-50 Modified, and EA-65 are alcohol-based stains used in the cytology laboratory for routine cytoplasmic staining using thePapanicolaou Method.
Orange G-6 is an alcohol-based stain used in the cytology laboratory to detect the presence of keratin in the cytoplasm. This formula is pre filtered before bottling and is manufactured for optimal consistency.
For routine cytoplasmic staining using Papanicolaou Method
Specifications
Type OG-6 Cytology Stain
For Use With (Application) Detect the presence of keratin in the cytoplasm
Quantity 3800mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.